Learn More
Köttermann Underbench Chemical Storage Cabinet
This lockable Underbench Chemical Storage Cabinet provides safe storage of aggressive non-flammable chemicals and toxic substances.
Brand: Köttermann 375-0005
Additional Details : Weight : 106.29230kg
Description
- For the storage of non-corrosive, non-flammable substances
- Can be integrated in modular cabinet assemblies
- Lockable against unauthorized access
- Capable of connecting to the exhaust air system for permanent ventilation
- Double-walled, noise-insulated doors
Specifications
516 x 900 x 1920 mm | |
2 Doors, 4 Shelves | |
Hinged, Locking |
White | |
Non-corrosive, Non-flammable Chemical Storage | |
Steel (Galvanized) |