missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNG Monoclonal Antibody (OTI2C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503563S
This item is not returnable.
View return policy
Description
UNG Monoclonal antibody specifically detects UNG in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| UNG | |
| Monoclonal | |
| 0.42 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NR50 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8891 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C12 | |
| Unconjugated | |
| EIF2B3 | |
| DGU; DKFZp781L1143; HIGM4; HIGM5; UDG; UNG; Ung1; UNG15; UNG2; unga; uracil DNA glycosylase; uracil DNA glycosylase a; uracil-DNA glycosylase; uracil-DNA glycosylase 1, uracil-DNA glycosylase 2; uracil-DNA glycosylase isoform UNG1; uracil-DNA glycosylase isoform UNG2; uracil-DNA glycosylase nuclear isoform; zgc:56102 | |
| Full length human recombinant protein of human EIF2B3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction