Learn More
Cytiva UNICORN™ Start 1.3
Product Code. 17517196 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
ÄKTA™ Start
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17517196 ÄKTA™ Start Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17517196 Supplier Cytiva Supplier No. 29704232
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
UNICORN™ start is AKTA start™ control software. Cytiva recently updated to version 1.3.
  • UNICORN™ start is ÄKTA™ start control software
    • Simple and flexible method creation using predefined templates and phases (steps)
    • Enables real-time monitoring of the run and an intuitive way to control the run
    • Evaluate, compare, and share results. Create and print PDF reports
    • Robust database storage with easy access to data, data security, and data integrity
  • UNICORN™ start software includes four different modules: System Control, Method Editor, Evaluation, and Administration, allowing you to design runs, operate the ÄKTA™ start instrument, and to evaluate and share results. While ÄKTA™ start can function as a stand-alone system, UNICORN™ start software maximizes the performance of the instrument.
  • Visualize your run
    • Monitor your run using the interactive process picture.
    • This valuable functionality gives you real-time information about the current step, chromatogram, and various curves; flow, gradient, pressure and conductivity curves are displayed, along with fractions.
  • Simple and flexible method creation
    • Methods can be created using templates like affinity, ion exchange, gel filtration (also known as size exclusion chromatography), and desalting.
    • Customized methods can be created simply by dragging and dropping chromatography phases such as Prime and Equilibration, Sample Application, Wash out Unbound, Elution and Fractionation from the phase library.
    • Methods can be run directly from the System Control module or simply transferred to a USB memory stick and imported into ÄKTA™ start if the system is being used as a stand-alone unit (i.e., the system is not connected to a computer).
  • Single-click operations
    • UNICORN™ start software has an intuitive graphic user interface that enables users to perform operations like controlling flow path, peak integration, and report generation in a single-click.
TRUSTED_SUSTAINABILITY
Specifications
Type Software
For Use With (Equipment) ÄKTA™ Start
Includes System Control, Method Editor, Evaluation, and Administration Modules