Learn More
Superfos™ UniPak™ Polypropylene Bucket
Provides flexible packaging. Superfos™ UniPak™ Polypropylene Buckets match each lid diameter so only one versatile type of lid is needed for a range of sizes. Tamper resistant.
Brand: Superfos™ 5030-WHITE
Additional Details : Weight : 0.03440kg
Description
- Protective collar
- Withstands hot filling and blast freezing
- Available in round, rectangular, oval and square shapes
- Broad range of standard sizes
- Liquid tight, snap-on lid
- Same lid fits several volumes
- Excellent decoration options with In-Mould Labelling
- Visible deterrent to tampering