Learn More
Alexandra™ Unisex Lab Coat with Hip Pockets, Royal Blue
Product Code. 15881124
Change view
Click to view available options
Size:
Medium
Large
X-Large
3X-Large
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15881124 X-Large Each
15811134 3X-Large Each
15851124 Medium Each
15861124 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15881124 Supplier Alexandra™ Supplier No. G178ROXL
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Unisex asymmetrical stud front laboratory coat. Features stretch cuffs for comfort and three useful pockets. Available in a choice of colors.
  • Two hip pockets one chest pocket
  • Machine washable and industrially launderable
  • Unisex
  • Length 43 in./ 110cm
  • Fabric Weight: 245gsm
  • 67% polyester/33% cotton
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Polycotton
Color Royal Blue
Disposable Reusable
Sterility Non-sterile
Model G178
Size (English) Chest 44 in.
Weight (Metric) 245 gsm