Learn More
Carl Roth™ Universal Absorber Rotisorb™
Description
Ideal for absorption of inflammable, oily, and aggressive liquids such as:
- anorganic and organic acids
- alkaline solutions
- alcohols
- aldehydes
- aminesesters
- aromatic, chlorinated and aliphatic hydrocarbons
- substance compounds, e. g. paint, heating oil, fuels, lacquers, motor lubricants
Specifications
Specifications
| Type | Universal Absorber |
| Packaging Quantity | 1 Ea. |