Learn More
Lenz Laborglasinstrumente™ Universal Connectors
Product Code. 10008450
Change view
Click to view available options
Diameter (Metric) Inner:
6 to 14 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10008450 6 to 14 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10008450 Supplier Lenz Laborglasinstrumente™ Supplier No. 01601000
  • ONLINE EXCLUSIVE PRICE
    € 8.00
    € 5.73 / Pack of 1
    Save € 2.27
    28% Off
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Made of DURAN™ tubing. Lenz Laborglasinstrumente™ Universal Connectors are for use with connecting hoses with different diameters.
  • Made of DURAN™tubing
  • Universal connector
Specifications
Diameter (Metric) Inner 6 to 14 mm
Overall Length (Metric) 110 mm