Learn More
R&D Systems™ Universal Kinase Activity Kit, 2 Plate
Product Code. 18741864
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18741864 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18741864 Supplier R&D Systems™ Supplier No. EA004
Includes
CHO cell-expressed Recombinant Mouse CD39L2/ENTPD6, ATP, ADP, Phosphatase Buffer 4, Phosphate Standard, Malachite Green Reagents A and B
A simple, nonradioactive high throughput-compatible format for assaying the enzyme activity of kinases in vitro
The majority of kinases use ATP as the phosphate donor. They transfer the terminal phosphate group of ATP to a substrate, producing ADP as a by-product.
Kit is an ADP-based phosphatase-coupled kinase assay that ues CD39L2/ENTPD6 as a coupling phosphatase
CD39L2 releases the beta-phosphate from ADP, and the released inorganic phosphate is detected using malachite green reagents
The amount of inorganic ph
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit