missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Universal Sterile Filter Tips for Gilson™ P1000
Description
- Minimizes danger of unwanted cross-contamination
- Acts as reliable barrier against the transfer or aerosols into shaft of pipette
- Can be used with all standard pipettes
- Maintain tolerance limits stipulated for pipette tips i
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | White |
| Sterilization Method | Irradiation |
| For Use With (Equipment) | Anthos New Line, Costar Onepette, Eppendorf Research, Thermo Scientific Finnpipette Digital, Gilson Pipetman, Gilson Pipetman P200, Human Pipettes |
| Graduated | Yes |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?