Learn More
Kern Sohn Unlined Wooden Box for OIML Class F2 and M3 Weights
Click to view available options
Holds:
1 mg to 1 kg
1 mg to 10 kg
1 mg to 2 kg
1 mg to 200 g
1 mg to 500 g
1 mg to 6 kg
Unit Size:
Each
Description
- Wooden box for tailor made weight set
- For weight classes F2, M1, M2 and M3
Specifications
Specifications
| Description | Unlined |
| Material | Wood |
| For Use With (Equipment) | Class F2, M1, M2 and M3 Weights |
| Holds | 1 mg to 200 g |