Learn More
Macherey-Nagel™ Untreated Capillary
Product Code. 10688611 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
1 m
10 m
25 m
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Pack of 10
Pack of 3
Product Code. 10688611
Brand: Macherey-Nagel™ 723501.10
Only null left
Specifications
Diameter (Metric) 0.8 mm
Type Untreated Capillary
Diameter (Metric) Inner 0.53 mm
For Use With (Application) Preparation of Capillary Columns Analysis, Preparation of Capillary Columns, LC
Length (Metric) 10 m
Quantity 1 Each