missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UoS 12258
GREENER_CHOICE

Product Code. 18799446
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18799446 10 mg 10mg
18797836 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18799446 Supplier Tocris Bioscience™ Supplier No. 6353/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of AMPA receptors (pEC50 = 5.2 for potentiation of AMPA-mediated currents in vitro)

UoS 12258 is a positive allosteric modulator of AMPA receptors (pEC50 = 5.2 for potentiation of AMPA-mediated currents in vitro). Also potentiates AMPA-mediated synaptic responses in vivo. Improves performance of rats in various learning and memory paradigms
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(2S)-5-(6-Fluoro-3-pyridinyl)-2,3-dihydro-1H-inden-2-yl]-2-propanesulfonamide
CAS 875927-64-3
Quantity 10 mg
Molecular Formula C17H19FN2O2S
Formula Weight 334.41
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.