missing translation for 'onlineSavingsMsg'
Learn More

Upadacitinib, MedChemExpress

Product Code. 16411906
Encompass
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
25mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16411906

Brand: Medchem Express HY19569

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relevant cytokines. Upadacitinib (ABT-494) can be used for several autoimmune disorders research.

Upadacitinib (ABT-494) is a potent, orally active and selective Janus kinase 1 (JAK1) inhibitor (IC50=43 nM). Upadacitinib (ABT-494) displays approximately 74 fold selective for JAK1 over JAK2 (200 nM) in cellular assays dependent on specific, relevant cytokines. Upadacitinib (ABT-494) can be used for several autoimmune disorders research.

Specifications

Chemical Name or Material Upadacitinib
Physical Form Powder
CAS 1310726-60-3
Color Gray
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C17H19F3N6O
For Use With (Application) COVID-19-immunoregulation
Synonym ABT-494
Solubility Information DMSO : ≥ 22 mg/mL (57.84 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N1C[C@@H](CC)[C@@H](C2=CN=C3C=NC(NC=C4)=C4N32)C1)NCC(F)(F)F
Molecular Weight (g/mol) 380.37
Formula Weight 380.37
Percent Purity 98.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.