missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (109801) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB531JF525
This item is not returnable.
View return policy
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Flow Cytometry,Western Blot,Blockade of Receptor-ligand Interaction,CyTOF-reported
Specifications
| uPAR | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1, Leu24-Thr297, Accession # P35456 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Western Blot, Blockade of Receptor-ligand Interaction, CyTOF | |
| 109801 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cancer | |
| 5329 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction