Learn More
uPAR Antibody (109801), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB531MFV610
Product Code. 18599114
  • € 718.00 / 0.10mL
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blocking Assay, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
uPAR
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Rat
0.1 mL
Cancer
5329
Store at 4C in the dark.
Western Blot, Flow Cytometry, Blocking Assay, CyTOF
109801
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-reported
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3
Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1, Leu24-Thr297, Accession # P35456