missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (109801), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB531PEATT594
This item is not returnable.
View return policy
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Flow Cytometry
Specifications
| uPAR | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 109801 | |
| Flow Cytometry | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1, Leu24-Thr297, Accession # P35456 | |
| 0.1 mL | |
| Cancer | |
| 5329 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction