Learn More
uPAR Antibody (109801), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB531PECY55
Product Code. 30027801
  • € 729.00 / 0.10mL
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
uPAR
Monoclonal
PE-Cyanine5.5
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry
109801
Flow Cytometry
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3
Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1, Leu24-Thr297, Accession # P35456
0.1 mL
Cancer
5329
Store at 4C in the dark. Do not freeze.