missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 301.00 - € 461.00
Specifications
| Antigen | uPAR |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18405811
|
Novus Biologicals
NBP2-14711-25ul |
25ul |
€ 301.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18196421
|
Novus Biologicals
NBP2-14711 |
0.1 mL |
€ 461.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
uPAR Polyclonal specifically detects uPAR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| uPAR | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5329 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: VRLWEEGEELELVEKSCTHSEKTNRTLSYRTGLKITSLTEVVCGLDLCNQGNSGRAVTYSRSRYLECISCGSSDMSCERGRHQSLQCRSPEEQCLDVVTHWIQEGE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| PLAUR | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title