missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (RM0143-7J34), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22519DL594
This item is not returnable.
View return policy
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Western Blot
Specifications
| uPAR | |
| Monoclonal | |
| DyLight 594 | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Mouse recombinant protein of uPAR | |
| 0.1 mL | |
| Cancer | |
| 5329 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0143-7J34 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction