missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Rabbit anti-Mouse, Clone: 14, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 541.00
Specifications
| Antigen | uPAR |
|---|---|
| Clone | 14 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203698
|
Novus Biologicals
NBP2-90454-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18214727
|
Novus Biologicals
NBP2-90454-100UL |
100 μg |
€ 541.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
uPAR Monoclonal specifically detects uPAR in Mouse samples. It is validated for ELISA.Specifications
| uPAR | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Recombinant Mouse uPAR Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 14 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 5329 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title