missing translation for 'onlineSavingsMsg'
Learn More

Idex Bottle Caps

Product Code. p-4910502
Click to view available options
For Use With (Equipment):
0.06 in. O.D. tubing
Thread Size (Metric):
GL 38
GL 45
Color:
Aqua Blue
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12905600

Brand: Idex 0203335

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Bottle caps used for GL-38 and GL-45 tubing

Specifications

Color Aqua Blue
Material Polypropylene
No. per Case 1 Cs.
Thread Size (Metric) GL 38
Product Type Cap
For Use With (Equipment) 0.06 in. O.D. tubing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.