Learn More
Kinesis™ Peristaltic Tubing Adapters
Product Code. 13050410
Change view
Click to view available options
Diameter (English) Inner:
0.048 to 0.110 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
13050410 0.048 to 0.110 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13050410 Supplier Kinesis™ Supplier No. 0201439
Only null left
Add to basket
Add to basket
Connect 1/16 in. OD rigid/semi-rigid tubing to soft-wall tubing
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.
Specifications
Type Peristaltic Tubing Adapter
Diameter (English) Inner 0.048 to 0.110 in.
For Use With (Equipment) Peristaltic to 1/16 in. O.D. Rigid