Learn More
Idex High-Pressure Inline Filters
Prevent problems stemming from particulate matter and protect major system components by using these filters
Brand: Idex 02010-10
Additional Details : Weight : 0.25000kg
Description
Prevent problems stemming from particulate matter and protect major system components by using these filters
- Excellent choices for inline filtration needs
- Multiple versions customized for different applications available
Specifications
2 μm | |
PEEK™ | |
High-Pressure Inline Filter |