Learn More
Kinesis™ PEEK Chromatography Tubing
Click to view available options
Diameter (English) Inner:
0.005 in.
0.007 in.
0.01 in.
0.02 in.
0.03 in.
0.04 in.
Overall Length (English):
5 ft.
50 ft.
100 ft.
Maximum Pressure:
5000 psi (345 bar)
7000 psi
7000 psi (483 bar)
Color:
Blue
Green
Natural
Orange
Red
Yellow
Unit Size:
Each
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
- Biocompatible, inert and easy to cut
- Great for high-pressure applications
- Wide choice of outer and inner diameters
- Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
| Type | Tubing |
| Color | Natural |
| For Use With (Application) | High-Pressure Applications |
| Diameter (English) Inner | 0.04 in. |
| Overall Length (English) | 5 ft. |
| Material | PEEK |
| Maximum Pressure | 5000 psi (345 bar) |