missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94094-0.02ml
This item is not returnable.
View return policy
Description
UPF2 Polyclonal antibody specifically detects UPF2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| UPF2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| DKFZP434D222, hUpf2, KIAA1408MGC138835, Nonsense mRNA reducing factor 2, regulator of nonsense transcripts 2, RENT2MGC138834, smg-3, UPF2 regulator of nonsense transcripts homolog (yeast), Up-frameshift suppressor 2 homolog, yeast Upf2p homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1153-1272 of human UPF2 (NP_542166.1). GPPLGGGEGEAESADTMPFVMLTRKGNKQQFKILNVPMSSQLAANHWNQQQAEQEERMRMKKLTLDINERQEQEDYQEMLQSLAQRPAPANTNRERRPRYQHPKGAPNADLIFKTGGRRR | |
| 0.02 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26019 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction