missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 493.00
Specifications
| Antigen | UPF2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625540
|
Novus Biologicals
NBP2-94519-0.02ml |
0.02 mL |
€ 216.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672761
|
Novus Biologicals
NBP2-94519-0.1ml |
0.1 mL |
€ 493.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UPF2 Polyclonal antibody specifically detects UPF2 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| UPF2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| DKFZP434D222, hUpf2, KIAA1408MGC138835, Nonsense mRNA reducing factor 2, regulator of nonsense transcripts 2, RENT2MGC138834, smg-3, UPF2 regulator of nonsense transcripts homolog (yeast), Up-frameshift suppressor 2 homolog, yeast Upf2p homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1153-1272 of human UPF2 (NP_542166.1). GPPLGGGEGEAESADTMPFVMLTRKGNKQQFKILNVPMSSQLAANHWNQQQAEQEERMRMKKLTLDINERQEQEDYQEMLQSLAQRPAPANTNRERRPRYQHPKGAPNADLIFKTGGRRR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 26019 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title