missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | UPF2 |
|---|---|
| Concentration | 260 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233490
|
Proteintech
55022-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234227
|
Proteintech
55022-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UPF2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human UPF2. This antibody recognizes human antigen. The UPF2 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| UPF2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp434D222; HUPF2; KIAA1408; MGC138834; MGC138835; RENT2; smg-3 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 260 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 26019 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title