missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPF3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584488
This item is not returnable.
View return policy
Description
Immunogen sequence: FLETYCVEEE KTSANPETLL GEMEAKTREL IARRTTPLLE YIKNRKLEKQ R Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 88%.
UPF3A is part of a multiprotein post-splicing mRNP complex involved in both mRNA nuclear export and mRNA surveillance. UPF3A is involved in nonsense-mediated decay (NMD) of mRNAs containing premature stop codons.
Specifications
| UPF3A | |
| Polyclonal | |
| Unconjugated | |
| UPF3A | |
| 2600001C03Rik; 4930546M19Rik; hUpf3; HUPF3A; nonsense mRNA reducing factor 3A; Regulator of nonsense transcripts 3A; RENT3A; UPF3; UPF3 regulator of nonsense transcripts homolog A (yeast); UPF3A; Up-frameshift suppressor 3 homolog A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H1J1 | |
| UPF3A | |
| Recombinant protein corresponding to Human UPF3A. Recombinant protein control fragment (Product #RP-107052). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction