missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UPK1B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15734771 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15734771 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15734771 Supplier Invitrogen™ Supplier No. PA556863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FTPNLFLKQM LERYQNNSPP NNDDQWKNNG VTKTWDRLML QDNCCGVNGP SDW Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.

Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPK1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UPK1B
Gene Accession No. O75841
Gene Alias AI413235; tetraspan; tetraspanin-20; Tspan20; Tspan-20; UP1b; UPIb; Upk1; Upk1b; uroplakin 1B; uroplakin Ib; Uroplakin-1b
Gene Symbols UPK1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UPK1B. Recombinant protein control fragment (Product #RP-95613).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7348
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.