missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560318
This item is not returnable.
View return policy
Description
Immunogen sequence: ISSLSGLLSP ALTESLLVAL PPCHLTGGNA TLMVRRANDS KVVTSSFVVP PCRGRRELVS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
This gene encodes one of the proteins of the highly conserved urothelium-specific integral membrane proteins of the asymmetric unit membrane which forms urothelium apical plaques in mammals. The asymmetric unit membrane is believed to strengthen the urothelium by preventing cell rupture during bladder distention. The encoded protein is expressed in the peripheral blood of bladder cancer patients with transitional cell carcinomas.
Specifications
| UPK2 | |
| Polyclonal | |
| Unconjugated | |
| UPK2 | |
| AW491716; UP2; UPII; UPK2; uroplakin 2; uroplakin II; Uroplakin-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00526 | |
| UPK2 | |
| Recombinant protein corresponding to Human UPK2. Recombinant protein control fragment (Product #RP-98390). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction