missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPK3A Recombinant Rabbit Monoclonal Antibody (PD00-09)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544649
This item is not returnable.
View return policy
Description
UPK3A Recombinant Monoclonal Antibody for IHC (P), Flow
This gene encodes a member of the uroplakin family, a group of transmembrane proteins that form complexes on the apical surface of the bladder epithelium. Mutations in this gene may be associated with renal adysplasia. Alternatively spliced transcript variants have been described.
Specifications
| UPK3A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75631 | |
| Upk3a | |
| Synthetic peptide within Human Uroplakin III aa 250 to the C-terminus. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PD00-09 | |
| Unconjugated | |
| Upk3a | |
| 1110017C07Rik; proplakin IIIa; UP3a; UPIII; UPIIIA; Upk3; UPK3A; uroplakin 3; uroplakin 3A; uroplakin III; Uroplakin-3a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7380 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction