missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPP2 Monoclonal Antibody (OTI10A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812317
This item is not returnable.
View return policy
Description
UPP2 Monoclonal antibody specifically detects UPP2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| UPP2 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P97576, Q99LP6, Q9HAV7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 17713, 79563, 80273 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI10A3 | |
| Unconjugated | |
| GRPEL1 | |
| 1700124F02Rik; AI266885; liver-specific uridine phosphorylase; L-UrdPase; UDRPASE2; UP2; UPase 2; UPASE2; UPP2; urdPase 2; uridine phosphorylase 2; uridine phosphorylase-2 | |
| Human recombinant protein fragment corresponding to amino acids 28-217 of human GRPEL1 (NP_079472) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction