missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPRT Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506869S
This item is not returnable.
View return policy
Description
UPRT Monoclonal antibody specifically detects UPRT in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| UPRT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96BW1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 139596 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C9 | |
| Unconjugated | |
| UPRT | |
| FUR1; Gm774; RP11-311P8.3; UMP pyrophosphorylase; UPP; Uprt; UPRTase; uracil phosphoribosyltransferase (FUR1) homolog; uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae); uracil phosphoribosyltransferase homolog | |
| Full length human recombinant protein of human UPRT produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction