Learn More
Chicken anti-Rabbit IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75265PCP
Product Code. 30228185
  • ONLINE EXCLUSIVE PRICE
    € 628.00
    € 593.25 / 1mL
    Save € 34.75
    5% Off
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
PBS
This Chicken anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rabbit IgG (H&L).
1 mL
Adaptive Immunity, Immunology
Rabbit
Purified
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay
PerCP
Chicken
Affinity purified
RUO
Secondary
Store at 4°C in the dark.