missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRC1 Antibody (OTI1G6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74796
This item is not returnable.
View return policy
Description
UQCRC1 Monoclonal specifically detects UQCRC1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| UQCRC1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready | |
| Complex III subunit 1, Core protein I, cytochrome b-c1 complex subunit 1, mitochondrial, D3S3191, QCR1, ubiquinol-cytochrome c reductase core protein I, Ubiquinol-cytochrome-c reductase complex core protein 1, UQCR1 | |
| Mouse | |
| 52.5 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| OTI1G6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| UQCRC1 | |
| Human recombinant protein fragment corresponding to amino acids 46-305 of human UQCRC1 (NP_003356) produced in E.coli. | |
| Affinity Purified | |
| metabolism | |
| 7384 | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction