Learn More
UQCRFS1 Mouse anti-Human, Mouse, Rat, Clone: OTI4H8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811461
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.Specifications
| UQCRFS1 | |
| Monoclonal | |
| Unconjugated | |
| UQCRFS1 | |
| RIP1, RIS1, RISP, UQCR5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 291103, 66694, 7386 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P20788, P47985, Q9CR68 | |
| Uqcrfs1 | |
| Human recombit protein fragment corresponding to amino acids 79-274 of human UQCRFS1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.