Specifications
| Timer | Yes |
| Frequency | 50/60 Hz |
| Voltage | 230 V |
| Capacity (Metric) | 5.7 L |
| Electrical Requirements | 230V, 50/60Hz |
| For Use With (Application) | Cleaning test sieves and grinding tools quickly and easily |
| Height (Metric) Exterior | 13 cm |
| Model | UR 1 |