missing translation for 'onlineSavingsMsg'
Learn More

URB1 Antibody - BSA Free, Novus Biologicals™

Product Code. 30558778 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558778 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558778 Supplier Novus Biologicals Supplier No. NBP345634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

URB1 Polyclonal antibody specifically detects URB1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen URB1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias C21orf108, chromosome 21 open reading frame 108, KIAA0539nucleolar pre-ribosomal-associated protein 1, NPA1nucleolar preribosomal-associated protein 1, URB1 ribosome biogenesis 1 homolog, URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen URB1 ribosome biogenesis 1 homolog. (Uniprot# O60287)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9875
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.