Learn More
URG4 Mouse anti-Human, Unconjugated, Clone: 2D5F12, Proteintech
Product Code. 16889605 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16889605 20 μL 20µL
16879605 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16889605 Supplier Proteintech Supplier No. 671441IG20UL
Mouse Monoclonal Antibody
The URG4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human URG4. This antibody recognizes human, mouse, rat antigen. The URG4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Antigen URG4
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 2D5F12
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 55665
Target Species Human, Rat, Mouse
Isotype IgG2a