missing translation for 'onlineSavingsMsg'
Learn More
Learn More
URI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59777
This item is not returnable.
View return policy
Description
URI Polyclonal specifically detects URI in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| URI | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| chromosome 19 open reading frame 2, FLJ10575, NNX3RPB5-mediating protein, Protein NNX3, RMPunconventional prefoldin RPB5 interactor, RNA polymerase II subunit 5-mediating protein, URIRNA polymerase II, subunit 5-mediating protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8725 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| BAA34781.1 | |
| URI1 | |
| The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human RNA polymerase II, subunit 5 (RPB5)-mediating protein using the numbering given in entry BAA34781.1 (GeneID 8725). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only
Spot an opportunity for improvement?