missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UROC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584829
This item is not returnable.
View return policy
Description
Immunogen sequence: TDSPFRETSN IYDGSAFCAD MAVQNFVGDA CRGATWVALH NGGGVGWGEV INGGFGLVLD GTPEAEGRAR LMLSWDVSNG V Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 95%.
This gene encodes an enzyme involved in histidine catabolism, metabolizing urocanic acid to formiminoglutamic acid. The gene product is known to protect the skin from ultra violet rays and is contained in human sweat. Deficiency of this gene product in the liver is an apparent cause of mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| UROC1 | |
| Polyclonal | |
| Unconjugated | |
| UROC1 | |
| AI265605; BC022133; HMFN0320; Imidazolonepropionate hydrolase; UROC1; Urocanase; urocanase 1; urocanase domain containing 1; urocanate hydratase; urocanate hydratase 1; UROCD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 131669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96N76 | |
| UROC1 | |
| Recombinant protein corresponding to Human UROC1. Recombinant protein control fragment (Product #RP-108246). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur