missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroguanylin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94611-0.02ml
This item is not returnable.
View return policy
Description
Uroguanylin Polyclonal antibody specifically detects Uroguanylin in Mouse samples. It is validated for Western Blot
Specifications
| Uroguanylin | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| GCAP-II, guanylate cyclase activator 2B, guanylate cyclase activator 2B (uroguanylin), UGN, uroguanylin | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GUCA2B (NP_009033.1). MGCRAASGLLPGVAVVLLLLLQSTQSVYIQYQGFRVQLESMKKLSDLEAQWAPSPRLQAQSLLPAVCHHPALPQDLQPVCASQEASSIFKTLRTIANDDC | |
| 0.02 mL | |
| Cardiovascular Biology, Cell Biology, Neuroscience, Signal Transduction | |
| 2981 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction