missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ia Antibody (UPK1A/2922), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79869JF549
This item is not returnable.
View return policy
Description
Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Uroplakin Ia | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UPK1A | |
| Recombinant fragment (around aa 114-173) of human Uroplakin Ia (UPK1A) protein (exact sequence is proprietary) (Uniprot: O00322) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| UPK1A/2922 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction