missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ib Antibody (UPK1B/3273), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79935H
This item is not returnable.
View return policy
Description
Uroplakin Ib Monoclonal specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Uroplakin Ib | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| UPK1B | |
| Recombinant fragment (around aa 109-229) of human Uroplakin Ib (UPK1B) protein (exact sequence is proprietary) (Uniprot: O75841) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| UPK1B/3273 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction