missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ib Antibody (UPK1B/9258R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24164F
This item is not returnable.
View return policy
Description
Uroplakin Ib Monoclonal antibody specifically detects Uroplakin Ib in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Uroplakin Ib | |
| Monoclonal | |
| FITC | |
| tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
| Recombinant fragment (around aa 109-229) of human Uroplakin Ib protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Signal Transduction | |
| 7348 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| UPK1B/9258R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction