missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin IIIB Antibody (UPK3B/3270), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20910AF532
This item is not returnable.
View return policy
Description
Uroplakin IIIB Monoclonal antibody specifically detects Uroplakin IIIB in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Uroplakin IIIB | |
| Monoclonal | |
| Alexa Fluor 532 | |
| FLJ32198, MGC10902, p35, UP3b, UPIIIB, UPIIIbP35, uroplakin 3B, Uroplakin IIIb, uroplakin-3b | |
| Recombinant fragment (around aa 100-300) of human Uroplakin IIIB protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 105375355 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| UPK3B/3270 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction