missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin IIIB Antibody (UPK3B/8550R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 298.00 - € 530.00
Specifications
| Antigen | Uroplakin IIIB |
|---|---|
| Clone | UPK3B/8550R |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612974
|
Novus Biologicals
NBP3-20602-100ug |
100 μg |
€ 530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683334
|
Novus Biologicals
NBP3-20602-20ug |
20 μg |
€ 298.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uroplakin IIIB Monoclonal antibody specifically detects Uroplakin IIIB in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| Uroplakin IIIB | |
| 0.2 mg/ml | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10mM PBS with 0.05% BSA | |
| 105375355 | |
| IgG κ | |
| Protein A or G purified |
| UPK3B/8550R | |
| ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ32198, MGC10902, p35, UP3b, UPIIIB, UPIIIbP35, uroplakin 3B, Uroplakin IIIb, uroplakin-3b | |
| Recombinant fragment (around aa 100-300) of human Uroplakin IIIB protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title