missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UROS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528322
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Pig (88%), Rhesus Monkey (97%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene catalyzes the fourth step of porphyrin biosynthesis in the heme biosynthetic pathway. Defects in this gene cause congenital erythropoietic porphyria (Gunther's disease).
Specifications
| UROS | |
| Polyclonal | |
| Unconjugated | |
| Uros | |
| AI415298; hydroxymethylbilane hydrolyase; Hydroxymethylbilane hydrolyase [cyclizing]; urogen III synthase; UROIIIS; uroporphyrinogen III co-synthase; uroporphyrinogen III synthase; uroporphyrinogen-III cosynthase; Uroporphyrinogen-III synthase; UROS; Uros3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7390 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P10746 | |
| Uros | |
| Recombinant protein encompassing a sequence within the center region of human UROS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction