missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urotensin-II R/GPR14 Antibody (443707) [DyLight 680], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9245Y
This item is not returnable.
View return policy
Description
Urotensin-II R/GPR14 Monoclonal antibody specifically detects Urotensin-II R/GPR14 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Urotensin-II R/GPR14 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 443707 | |
| Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 14, GPR14, G-protein coupled receptor 14, UR-2-R, UR-II-R, urotensin 2 receptor, urotensin II receptor, urotensin-2 receptor, UTR, UTR2 | |
| Mouse Urotensin-II R synthetic peptide | |
| 0.1 mL | |
| GPCR | |
| 2837 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction