missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Urotensin II Receptor Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534299

Product Code. 15305071

  • € 560.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%).

The Urotensin-II receptor has been reported to be expressed at high levels in the heart and pancreas and also in artery and smooth muscle. Low transcript levels have been identified in brain, particularly in thalamus, and superior occipital gyrus. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Urotensin II Receptor
Polyclonal
Unconjugated
UTS2R
G protein-coupled receptor 14; GPR14; G-protein coupled receptor 14; G-protein coupled sensory epithelial neuropeptide-like receptor; putative G protein-coupled receptor (SENR); Senr; Uiir; Uii-r; Uii-r1a; UII-receptor; UR2R; UR-2-R; UR-II-R; urotensin 2 receptor; urotensin II receptor; Urotensin-2 receptor; UT-II; UTIIR; UTR; UTR2; Uts2r
Rabbit
Antigen affinity chromatography
RUO
2837
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9UKP6
UTS2R
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human UTS2R / GPR14.
50 μg
Primary
Human, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.